


Naphthalene ≥98.0% (by GC)
: TCI
:91-20-3
:
:C₁₀H₈
:MFCD00001742
:128.17 g/mol
:14,06370
:218°C
:79…82 °C
:1,145 g/cm³ (20 °C)
:79 °C
:Ambient
:202-049-5
:1334
:4.1,III
:GHS07
:GHS08
:GHS09
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:79.0 to 83.0 deg-C
Frequently Bought Together