
Isonicotinaldehyde oxime ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:696-54-8
:
Formula:C₆H₆N₂O
MDL Number:MFCD00006446
:122.13 g/mol
EINECS:211-796-6
:GHS07
Specification Test Results
- Appearance:White to Light yellow to Light red powder to crystal
- Purity (GC):min. 95.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:131.0 to 135.0 deg-C
- Solubility in Methanol:very faint turbidity
Specification Test Results
Frequently Bought Together