
3-Methyl-1-phenyl-2-pyrazolin-5-one ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:89-25-8
:
Formula:C₁₀H₁₀N₂O
MDL Number:MFCD00003138
:174.2 g/mol
Merck Index:14,06713
EINECS:201-891-0
:GHS07
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:128.0 to 131.0 deg-C
- Solubility in N,N-DMF:almost transparency
Specification Test Results
Frequently Bought Together