

(S)-(-)-2,3-Epoxy-1-propanol ≥96.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:60456-23-7
:
Formula:C₃H₆O₂
MDL Number:MFCD00074874
:74.08 g/mol
Flash Pt:72°C
UN:2810
ADR:6.1,III
:GHS06
:GHS08
Storage Temperature:Freezer
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (GC):min. 96.0 %
- Optical purity (GC):min. 97.0 ee%
Specification Test Results
Related Information
Frequently Bought Together