Anthrone ≥98.0% (by GC)
Supplier: TCI
About this item
Synonyms: Anthracen-9(10H)-one
- CAS Number:90-44-8
- Formula:C₁₄H₁₀O
- MDL Number:MFCD00001187
- Molecular Weight:194.23 g/mol
- Merck Index:14,00691
- EINECS:201-994-0
2 Options Available Below
Product Details & Documents
Synonyms: Anthracen-9(10H)-one
- CAS Number:90-44-8
- Formula:C₁₄H₁₀O
- MDL Number:MFCD00001187
- Molecular Weight:194.23 g/mol
- Merck Index:14,00691
- EINECS:201-994-0
Documents
Certificates
Search by Lot NumberSpecifications for Product Family
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:154.0 to 160.0 deg-C
- Solubility in hot Toluene:almost transparency
- suitability for glucose-analysis:Abs min. 0.70 (625nm ) in the presence of D-Glucose (10 ppm).



