
Methylenecyclohexane ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:1192-37-6
:
Formula:C₇H₁₂
MDL Number:MFCD00001661
:96.17 g/mol
Boiling Pt:101°C
Melting Pt:–106.7 °C
Density:0,8074 g/cm³ (20 °C)
Flash Pt:–6°C
Storage Temperature:Refrigerator
EINECS:214-752-4
UN:3295
ADR:3,II
:GHS02
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 97.0 %
Specification Test Results
Frequently Bought Together