Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Cyclohexanone ≥99.0% (by GC)

Flammable
Toxic
Health
Supplier:   TCI
Cyclohexanone ≥99.0% (by GC)
Supplier:  TCI
Certificates
Cyclohexanone ≥99.0% (by GC)

2 Options Available Below

Product Details & Documents
Synonyms:  Pimelic Ketone
  • CAS Number:108-94-1
  • Formula:C₆H₁₀(=O)
  • MDL Number:MFCD00001625
  • Molecular Weight:98.14 g/mol
  • Merck Index:14,02726
  • Boiling Pt:157°C
  • Melting Pt:–31 °C
  • Density:0,9478 g/cm³ (20 °C)
  • Flash Pt:44°C
  • Storage Temperature:Ambient
  • EINECS:203-631-1
  • UN:1915
  • ADR:3,III
  • REACH:01-2119453616-35
Specifications for Product Family

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 99.0 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    25 mL
    Log in for availability
    Log in for availability
    1 * 25 mL
    1 * 25 mL - 132,00 kr
    132,00 kr
    1 * 25 mL
       132,00 kr

  • Glass bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 197,00 kr
    197,00 kr
    1 * 500 mL
       197,00 kr