Order Entry
Denmark
ContactUsLinkComponent
2,6-Di-tert-butyl-p-cresol ≥99.0% (by GC)

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    White or Colorless powder to crystaline
  • Purity (GC):
    min. 99.0 %
  • Melting point:
    69.0 to 72.0 deg-C
  • Solubility in Methanol:
    almost transparency

Specification Test Results

Frequently Bought Together