
N-Boc-L-prolinol ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:69610-40-8
:
Formula:C₁₀H₁₉NO₃
MDL Number:MFCD00066232
:201.27 g/mol
Boiling Pt:150°C
EINECS:239-848-3
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Optical purity (LC):min. 98.0 ee%
- Melting point:60.0 to 64.0 deg-C
- Specific rotation [a]20/D:-52.0 to -58.0 deg (C=5, MeOH)
Specification Test Results
Frequently Bought Together