N-Boc-D-prolinol ≥98.0% (by GC)
: TCI
:83435-58-9
:
:C₁₀H₁₉NO₃
:MFCD00040580
:201.3 g/mol
:GHS07
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Optical purity (LC):min. 98.0 ee%
- Melting point:58.0 to 63.0 deg-C
- Specific rotation [a]20/D:+52.0 to +57.0 deg (C=5, MeOH)
Frequently Bought Together