Order Entry
Denmark
ContactUsLinkComponent
(1R,2S)-(-)-2-Amino-1,2-diphenylethanol ≥99.0% (by GC, titration analysis)
Irritant
(1R,2S)-(-)-2-Amino-1,2-diphenylethanol ≥99.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
undefined
Irritant
(1R,2S)-(-)-2-Amino-1,2-diphenylethanol ≥99.0% (by GC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (GC):
    min. 99.0 %
  • Purity (Nonaqueous Titration):
    min. 99.0 %
  • Melting point:
    143.0 to 146.0 deg-C
  • Specific rotation [a]20/D:
    -6.0 to -8.0 deg (C=0.6, EtOH)

Specification Test Results

Related Information

Frequently Bought Together