Galactitol ≥98.0% (by GC)
Leverandør: TCI
Certifikater
Om denne vare
CAS nummer:608-66-2
:
Formel:HOH₂C(CH(OH))₄CH₂OH
MDL nummer:MFCD00064288
:182.17 g/mol
Merck Index:14,04332
Opbevaringstemperatur:Ambient
EINECS:210-165-2
Specifikationstestresultater
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:186.0 to 191.0 deg-C
- Solubility in hot Water:almost transparency
Specifikationstestresultater
Frequently Bought Together