
2-Fluoro-4-nitrotoluene ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:1427-07-2
:
Formula:C₇H₆FNO₂
MDL Number:MFCD00007199
:155.13 g/mol
Boiling Pt:235 °C (1013 hPa)
Melting Pt:35…36 °C
Storage Temperature:Ambient
EINECS:215-845-2
ADR:6.1,III
:GHS07
Specification Test Results
- Appearance:White or Colorless to Light yellow powder to lump to clear liquid
- Purity (GC):min. 97.0 %
- Freezing point:32.0 to 35.0 deg-C
- Solubility in Methanol:very faint turbidity
Specification Test Results
Frequently Bought Together