Acetophenone ≥98.5% (by GC)
Supplier: TCI
Certificates
About this item
Synonyms: Methyl phenyl ketone, Phenyl methyl ketone
CAS Number:98-86-2
:
Formula:C₈H₈O
MDL Number:MFCD00008724
Boiling Pt:201.7
Molecular Weight:120.15 g/mol
Melting Pt:20
Flash Pt:80°C
Merck Index:14,00073
EINECS:202-708-7
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.5 %
Specification Test Results
Frequently Bought Together