Order Entry
Denmark
ContactUsLinkComponent
 
3-Methyl-1-phenyl-2-butanone ≥98.0% (by GC)
 :  TCI
 
undefined
3-Methyl-1-phenyl-2-butanone ≥98.0% (by GC)
 :  TCI

 

  • Appearance:
    Colorless to Yellow to Green clear liquid
  • Purity (GC):
    min. 98.0 %

 

Frequently Bought Together