
2,3-Difluoroaniline ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:4519-40-8
:
Formula:C₆H₅F₂N
MDL Number:MFCD00010298
:129.11 g/mol
Boiling Pt:69°C
Density:1,274 g/cm³ (20 °C)
Flash Pt:71°C
EINECS:224-847-2
UN:2810
ADR:6.1,III
:GHS06
Specification Test Results
- Appearance:Colorless to Brown clear liquid
- Purity (GC):min. 97.0 %
Specification Test Results
Frequently Bought Together