
2,4,5-Trifluorobenzoic acid ≥97.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:446-17-3
:
Formula:C₇H₃F₃O₂
MDL Number:MFCD00013306
:176.09 g/mol
Boiling Pt:237 °C (1013 hPa)
Melting Pt:99…101 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 97.0 %
- Purity (Neutralization titration):min. 97.0 %
- Melting point:95.0 to 100.0 deg-C
Specification Test Results
Frequently Bought Together