
5-Chlorosalicylic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:321-14-2
:
Formula:C₇H₅ClO₃
MDL Number:MFCD00002457
:172.57 g/mol
Boiling Pt:321 °C (1013 hPa)
Melting Pt:171…175 °C
Storage Temperature:Ambient
EINECS:206-283-9
UN:2811
ADR:6.1,III
:GHS06
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:172.0 to 178.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together