
4-Chloro-3-nitrobenzaldehyde ≥95.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:16588-34-4
:
Formula:C₇H₄ClNO₃
MDL Number:MFCD00007078
:185.57 g/mol
Melting Pt:57…60 °C
EINECS:240-645-7
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to Light red to Green powder to crystal
- Purity (Oxime Formation):min. 95.0 %
- Melting point:60.0 to 63.0 deg-C
Specification Test Results
Frequently Bought Together