
3,4-Dimethoxybenzoic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:93-07-2
:
Formula:C₉H₁₀O₄
MDL Number:MFCD00002500
:182.18 g/mol
Merck Index:14,09953
Boiling Pt:303 °C (1013 hPa)
Melting Pt:117…119 °C
Density:1,436 g/cm³ (-100 °C)
Flash Pt:≥ 100 °C
EINECS:202-215-7
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:180.0 to 184.0 deg-C
- Solubility in hot Methanol:almost transparency
Specification Test Results
Frequently Bought Together