Ethyl benzoate ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
Synonyms: Benzoic acid ethyl ester
CAS Number:93-89-0
:
Formula:C₉H₁₀O₂
MDL Number:MFCD00009109
Boiling Pt:212°C
Molecular Weight:150.18 g/mol
Melting Pt:-35
Flash Pt:99°C
Merck Index:14,03766
EINECS:202-284-3
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Colorless clear liquid
- Purity (GC):min. 99.0 %
- Acid Value:max. 0.2
Specification Test Results
Frequently Bought Together