
4-Hydroxybenzonitrile ≥98.0% (by GC, titration analysis)
: TCI
:767-00-0
:
:C₇H₅NO
:MFCD00002312
:146°C
:109…112 °C
:~ 1 g/cm³ (20 °C)
:119.12 g/mol
:Ambient
:212-175-2
:GHS07
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:111.0 to 115.0 deg-C
Frequently Bought Together