1,2-Dibromo-3,5-difluorobenzene ≥98.0% (by GC)
Catalog # TCIAD2023-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Light yellow to Dark green powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:36.0 to 39.0 deg-C
Specification Test Results
About this item
:
:271.89 g/mol
Boiling Pt:208 °C (1013 hPa)
Melting Pt:35…38 °C
Density:2,128 g/cm³ (20 °C)
:GHS07
Storage Temperature:Refrigerator
CAS Number:139215-43-3
Formula:C₆H₂Br₂F₂
MDL Number:MFCD00042559