1,8-Dibromohexadecafluorooctane ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:812-58-8
:
Formula:C₈Br₂F₁₆
MDL Number:MFCD00153111
:559.87 g/mol
Specification Test Results
- Appearance:White to Almost white powder to lump
- Purity (GC):min. 97.0 %
- Melting point:37.0 to 41.0 deg-C
Specification Test Results
Frequently Bought Together