



3-Methylpentane ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:96-14-0
:
Formula:C₆H₁₄
MDL Number:MFCD00009342
:86.18 g/mol
Boiling Pt:63°C
Melting Pt:–118°C
Density:0,664 g/cm³ (20 °C)
Flash Pt:–7°C
EINECS:202-481-4
UN:1208
ADR:3,II
:GHS02
:GHS07
:GHS08
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 99.0 %
Specification Test Results
Frequently Bought Together