



2-Methylpentane ≥98.0% (by GC)
: TCI
:107-83-5
:
:C₆H₁₄
:MFCD00009406
:60°C
:–154 °C
:0,653 g/cm³ (20 °C)
:–20°C
:Ambient
:203-523-4
:3,II
:86.18 g/mol
:1208
:GHS02
:GHS07
:GHS08
:GHS09
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
Frequently Bought Together