2-Methylhexane ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:591-76-4
:
Formula:C₇H₁₆
MDL Number:MFCD00009487
:100.2 g/mol
Boiling Pt:90°C
Melting Pt:–118 °C
Density:0,6793 g/cm³ (20 °C)
EINECS:209-730-6
UN:3295
ADR:3,II
:GHS02
:GHS07
:GHS08
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 99.0 %
Specification Test Results
Frequently Bought Together