Order Entry
Denmark
ContactUsLinkComponent
2-Methylbutane ≥97.0% (by GC)
Flammable
Irritant
Health
Environment
2-Methylbutane ≥97.0% (by GC)
Supplier:  TCI
Certificates
undefined
Flammable
Irritant
Health
Environment
2-Methylbutane ≥97.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless clear liquid
  • Purity (GC):
    min. 97.0 %

Specification Test Results

Related Information

Frequently Bought Together