2-Methyl-3-pentanone ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:565-69-5
:
Formula:C₆H₁₂O
MDL Number:MFCD00009314
:100.16 g/mol
Boiling Pt:115°C
Melting Pt:–69 °C
Density:0,8845 g/cm³ (20 °C)
Flash Pt:11°C
Storage Temperature:Ambient
EINECS:209-288-4
UN:1224
ADR:3,II
:GHS02
Specification Test Results
- Appearance:Colorless to Yellow to Green clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together