
1,3-Propanediol ≥98.0% (by GC)
: TCI
:504-63-2
:
:HO(CH₂)₃OH
:MFCD00002949
:76.1 g/mol
:14,09714
:214°C
:–32 °C
:1,0597 g/cm³ (20 °C)
:74°C
:207-997-3
:GHS07
:Ambient
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
Frequently Bought Together