Order Entry
Denmark
ContactUsLinkComponent
 
Ethylene glycol ≥99.5% (by GC)
 :  TCI
 
undefined
Ethylene glycol ≥99.5% (by GC)
 :  TCI

 

  • Environmentally Preferable:

 

 

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 99.5 %

 

Frequently Bought Together