Order Entry
Denmark
ContactUsLinkComponent
 
Digitoxin ≥97.0% (by HPLC)
Catalog # TCIAD0542-100MG
Supplier:  TCI
CAS Number:  
undefined
Digitoxin ≥97.0% (by HPLC)
Catalog # TCIAD0542-100MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 97.0 area%
  • Specific rotation [a]20/D:
    +16.0 to +18.5 deg (C=2, CHCl3 (stabilized with Ethanol))

Specification Test Results

About this item

 :
 :764.95 g/mol
Merck Index:14,03163
Melting Pt:257°C
EINECS:200-760-5
UN:2811
ADR:6.1,II
 :GHS06
 :GHS08
CAS Number:71-63-6
Formula:C₄₁H₆₄O₁₃
MDL Number:MFCD00003686