Order Entry
Denmark
ContactUsLinkComponent
 
2-Hydroxybenzimidazole ≥98.0% (by total nitrogen basis)
  TCIAH0739-25G
 :  TCI
 :  
undefined
2-Hydroxybenzimidazole ≥98.0% (by total nitrogen basis)
  TCIAH0739-25G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

 

 

  • Purity (with Total Nitrogen):
    min. 98.0 %

 

 

 :
 :134.14 g/mol
 :320…322 °C
 :1,42 g/cm³ (20 °C)
 :210-412-4
 :615-16-7
 :C₇H₆N₂O
 :MFCD00127894