Order Entry
Denmark
ContactUsLinkComponent
 
Irritant
4-(Trifluoromethoxy)benzenesulphonamide ≥98.0% (by HPLC, total nitrogen)
Catalog # TCIAT2427-5G
Supplier:  TCI
CAS Number:  
undefined
Irritant
4-(Trifluoromethoxy)benzenesulphonamide ≥98.0% (by HPLC, total nitrogen)
Catalog # TCIAT2427-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (with Total Nitrogen):
    min. 98.0 %
  • Melting point:
    147.0 to 151.0 deg-C

Specification Test Results

About this item

 :
 :241.19 g/mol
 :GHS07
CAS Number:1513-45-7
Formula:C₇H₆F₃NO₃S
MDL Number:MFCD00798381