
(R)-(-)-Carvone ≥99.0% (by GC)
: TCI
:6485-40-1
:
:C₁₀H₁₄O
:MFCD00001578
:150.22 g/mol
:14,01874
:227 °C (1013 hPa)
:25.2 °C
:0,9652 g/cm³ (15 °C)
:88°C
:229-352-5
:GHS07
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 99.0 %
- Specific rotation [a]20/D:-59.0 to -63.0 deg (neat)
Frequently Bought Together