Order Entry
Denmark
ContactUsLinkComponent
Acetonitrile ≥99.5% (by GC) for spectrophotometry
Flammable
Toxic
Health
Acetonitrile ≥99.5% (by GC) for spectrophotometry
Catalog # TCIAA0293-100ML
Supplier:  TCI
CAS Number:  
undefined
Flammable
Toxic
Health
Acetonitrile ≥99.5% (by GC) for spectrophotometry
Catalog # TCIAA0293-100ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Colorless clear liquid
  • Purity (GC):
    min. 99.5 %
  • Absorbance (210nm):
    max. 0.100
  • Absorbance (220nm):
    max. 0.020
  • Absorbance (230nm):
    max. 0.010

Specification Test Results

About this item

 :
 :41.05 g/mol
Merck Index:14,00070
Boiling Pt:82°C
Melting Pt:–46°C
Density:0,782 g/cm³ (20 °C)
Flash Pt:10°C (closed cup)
Storage Temperature:Ambient
EINECS:200-835-2
UN:1648
ADR:3,II
REACH:01-2119471307-38
 :GHS02
 :GHS06
 :GHS08
CAS Number:75-05-8
Formula:H₃CCN
MDL Number:MFCD00001878