Hexanal ≥98.0% (by GC)
: TCI
:66-25-1
:
:C₆H₁₂O
:MFCD00007027
:100.16 g/mol
:14,01760
:131°C
:–56 °C
:0,81 g/cm³ (20 °C)
:32°C
:Refrigerator
:200-624-5
:1207
:3,III
:GHS02
:GHS07
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
Frequently Bought Together