1-Bromo-2-chloroethane ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:107-04-0
:
Formula:BrH₂CCH₂Cl
MDL Number:MFCD00000962
:143.41 g/mol
Boiling Pt:106°C
Melting Pt:–17 °C
Density:1,7392 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:203-456-0
UN:2810
ADR:6.1,III
:GHS06
:GHS08
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together