Order Entry
Denmark
ContactUsLinkComponent
Triacetoxyethylsilane ≥96.0% (by GC)
 
Triacetoxyethylsilane ≥96.0% (by GC)
  TCIAE0525-25G
 :  TCI
 :  
undefined
 
Triacetoxyethylsilane ≥96.0% (by GC)
  TCIAE0525-25G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

 

 

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 96.0 %

 

 

 :
 :234.28 g/mol
 :109°C
 :106°C
 :241-677-4
 :1760
 :8,III
 :GHS05
 :17689-77-9
 :C₈H₁₄O₆Si
 :MFCD00053703