
N,N-Diethyl-m-toluamide ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:134-62-3
:
Formula:C₁₂H₁₇NO
MDL Number:MFCD00009046
:191.27 g/mol
Merck Index:14,02856
Boiling Pt:160°C
Melting Pt:–45 °C
Density:0,996 g/cm³ (20 °C)
Flash Pt:140°C
EINECS:205-149-7
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together