

Sudan I ≥93.0% (by HPLC)
TCIAP0585-25G
: TCI
:
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
- Appearance:Orange to Brown powder to crystal
- Purity (HPLC):min. 93.0 area%
- Lambda max.:477.0 to 483.0 nm (MeOH)
- Absorbance (E1%1cm):min. 570 (MeOH, 477.0 to 483.0 nm)
:
:248.28 g/mol
:444 °C (1013 hPa)
:135 °C
:1,119 g/cm³ (151 °C)
:212-668-2
:GHS07
:GHS08
:842-07-9
:C₁₆H₁₂N₂O
:MFCD00003911