Murexide
Supplier: TCI
About this item
Synonyms: Ammonium purpurate, Murexide triturate, Ammonium 5-(2,4,6-trioxoperhydropyrimidin-5-ylideneamino)barbiturate, Purpuric acid ammonium salt
- CAS Number:3051-09-0
- Formula:C₈H₈N₆O₆
- MDL Number:MFCD00012777
- Molecular Weight:284.19 g/mol
- Merck Index:14,06306
- Melting Pt:300 °C
- Storage Temperature:Ambient
- EINECS:221-266-6
3 Options Available Below
Product Details & Documents
Synonyms: Ammonium purpurate, Murexide triturate, Ammonium 5-(2,4,6-trioxoperhydropyrimidin-5-ylideneamino)barbiturate, Purpuric acid ammonium salt
- CAS Number:3051-09-0
- Formula:C₈H₈N₆O₆
- MDL Number:MFCD00012777
- Molecular Weight:284.19 g/mol
- Merck Index:14,06306
- Melting Pt:300 °C
- Storage Temperature:Ambient
- EINECS:221-266-6
Specifications for Product Family
Specification Test Results
- Appearance:Orange to Brown to Dark purple powder to crystal
- Lambda max.:520.0 to 524.0 nm (H2O)
- Absorbance (E1%1cm):min. 350 (H2O, 520.0 to 524.0 nm)
- Sensitiveness:to pass test



