Order Entry
Denmark
ContactUsLinkComponent
EGTA (ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid) ≥95.0% (by titrimetric analysis)
EGTA (ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid) ≥95.0% (by titrimetric analysis)
Catalog # TCIAE0106-25G
Supplier:  TCI
CAS Number:  
undefined
EGTA (ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid) ≥95.0% (by titrimetric analysis)
Catalog # TCIAE0106-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystaline
  • Purity (Chelometric Titration):
    min. 95.0 %
  • Solubility in 1mol/L NaOH:
    almost transparency

Specification Test Results

About this item

 :
Merck Index:14,03529
Melting Pt:241°C
 :380.35 g/mol
Storage Temperature:Ambient
EINECS:200-651-2
CAS Number:67-42-5
Formula:C₁₄H₂₄N₂O₁₀
MDL Number:MFCD00004291