Order Entry
Denmark
ContactUsLinkComponent
6-Nitro-o-toluonitrile ≥98.0% (by GC)
 
6-Nitro-o-toluonitrile ≥98.0% (by GC)
  TCIAM0867-5G
 :  TCI
 :  
undefined
 
6-Nitro-o-toluonitrile ≥98.0% (by GC)
  TCIAM0867-5G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

 

 

  • Appearance:
    Light orange to Yellow to Green powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    107.0 to 109.0 deg-C

 

 

 :
 :162.15
 :217-553-0
 :GHS07
 :1885-76-3
 :C₈H₆N₂O₂
 :MFCD00007266