L-Alanine ≥99.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:56-41-7
:
Formula:C₃H₇NO₂
MDL Number:MFCD00064410
:89.09 g/mol
Merck Index:14,00204
Boiling Pt:189 °C (1013 hPa)
Melting Pt:79 °C
Density:1,4 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-273-8
Specification Test Results
- Appearance:White powder to crystal
- Purity (Nonaqueous Titration):min. 99.0 % (after drying)
- Specific rotation [a]20/D:+14.0 to +15.0 deg (C=10, HCl) (after drying)
- Solubility in Water:almost transparency
Specification Test Results
Related Information
Frequently Bought Together