Order Entry
Denmark
ContactUsLinkComponent
 
L-Phenylalanine ≥98.0% (by HPLC, titration analysis)
 :  TCI
 
undefined
L-Phenylalanine ≥98.0% (by HPLC, titration analysis)
 :  TCI

 

  • Appearance:
    White powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Optical purity (LC):
    min. 99.0 ee%
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Solubility in hot Water:
    almost transparency

 

Frequently Bought Together