Sodium cholate ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:361-09-1
:206986-87-0
Formula:C₂₄H₃₉NaO₅
MDL Number:MFCD00150749
:430.56 g/mol
Melting Pt:198 °C
Storage Temperature:Ambient
EINECS:206-643-5
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 % (after drying)
- Specific rotation [a]20/D:+34.0 to +36.0 deg (C=0.6, EtOH)
- Solubility in Water:almost transparency
Specification Test Results
Frequently Bought Together