6-Methyluracil ≥99.0% (by HPLC, titration analysis)
: TCI
: 6-Methylpyrimidine-2,4(1H,3H)-dione, 4-Methyluracil, 6-Methyl-2,4(1H,3H)-pyrimidinedione
:626-48-2
:
:C₅H₆N₂O₂
:MFCD00006028
:14,06133
:358 °C (1013 hPa)
:117 °C
:126.11 g/mol
:210-949-4
:GHS08
- Environmentally Preferable:
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 99.0 area%
- Purity (Neutralization titration):min. 99.0 %
- Solubility in hot Water:almost transparency
Frequently Bought Together