Methyl-10-undecenoate ≥96.0% (by GC)
: TCI
:111-81-9
:
:C₁₂H₂₂O₂
:MFCD00016689
:198.31 g/mol
:248°C
:–27.5 °C
:0,889 g/cm³ (20 °C)
:122°C
:203-910-8
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 96.0 %
Frequently Bought Together