DL-Serine ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:302-84-1
:
Formula:C₃H₇NO₃
MDL Number:MFCD00064223
:105.09 g/mol
Merck Index:14,08460
Boiling Pt:395 °C (1013 hPa)
Melting Pt:243…244 °C
Density:1,543 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:206-130-6
Specification Test Results
- Appearance:White powder to crystal
- Purity (Nonaqueous Titration):min. 98.0 %
- Solubility in hot Water:almost transparency
Specification Test Results
Frequently Bought Together